Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45524

Supelco

Hexabromobenzene

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6Br6
CAS Number:
Molecular Weight:
551.49
Beilstein/REAXYS Number:
1912586
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

>300 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Brc1c(Br)c(Br)c(Br)c(Br)c1Br

InChI

1S/C6Br6/c7-1-2(8)4(10)6(12)5(11)3(1)9

InChI key

CAYGQBVSOZLICD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amina Salamova et al.
Environmental science & technology, 45(20), 8698-8706 (2011-09-29)
Air (vapor and particle) and precipitation samples were collected at five sites (two urban, one rural, and two remote) around the Great Lakes during 2005-2009 as a part of the Integrated Atmospheric Deposition Network (IADN). The concentrations of polybrominated diphenyl
Jiang-Ping Wu et al.
Environment international, 37(1), 210-215 (2010-10-19)
With the phaseout of brominated flame retardants (BFRs) polybrominated diphenyl ethers (PBDEs), some non-PBDE BFRs have prompted to be alternatives to the discontinued PBDEs. To assess the bioaccumulation potential of these chemicals, field bioaccumulation factors (BAFs) for several non-PBDE BFRs
Lewis T Gauthier et al.
Environmental science & technology, 43(2), 312-317 (2009-02-26)
The production and use of nonpolybrominated diphenyl ether (non-PBDE), brominated flame retardant (BFR) alternatives have been on the rise, although their assessment in environmental samples is largely understudied. In the present study, several non-PBDE BFRs were found in the egg
Silke Rupp et al.
Chemosphere, 60(11), 1644-1651 (2005-08-09)
Polybrominated diphenyl ethers (PBDEs) are a group of brominated flame retardants (BFRs) used mainly as additives in different kinds of plastic material. Various PBDEs are found in all environmental compartments as well as in tissue and blood serum of animals
Qiwu Zhang et al.
Chemosphere, 48(8), 787-793 (2002-09-12)
A hexabromobenzene (HBB, C6Br6) powder was subjected to co-grinding with excess CaO powder in air using a planetary ball mill to investigate mechanochemical debromination of the HBB sample. The reaction proceeds smoothly with an increase in both grinding time and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service