Cell biochemistry and function, 3(4), 267-272 (1985-10-01)
Effect of tricyclohexylhydroxytin (plictran) on Ca2+-ATPase activity was studied in rat brain synaptosomes under in vitro and in vivo conditions. Plictran inhibited basal Ca2+-ATPase activity with an IC50 value of 6 nM suggesting its interaction with calcium transport phenomenon. Plictran
Journal of biochemical toxicology, 2, 125-140 (1987-01-01)
Tricyclohexylhydroxytin, commonly known as Plictran, inhibited Na+, K+-ATPase activity of rat brain synaptosomes in a concentration-dependent manner with median inhibitory concentration (IC-50) of 2 microM. Both K+-stimulated para-nitrophenylphosphatase and [3-H]-ouabain binding to synaptosomes were also inhibited by Plictran with IC-50
[Determination of the active principle of a plictran preparation and its metabolites in environmental objects].
Journal of biochemical toxicology, 1(4), 55-66 (1986-12-01)
Previous studies from this laboratory have indicated that tricyclohexyltin hydroxide (Plictran) is a potent inhibitor of both basal- and isoproterenol-stimulated cardiac sarcoplasmic reticulum (SR) Ca2+-ATPase, with an estimated IC-50 of 2.5 X 10(-8) M. The present studies were initiated to
Cyanazine, cyhexatin, dicamba and DNOC are pesticides commonly and broadly used in agriculture pest control. However, there is little information on their toxicity and mutagenicity in human cells and in whole animals. Therefore, UDS assay and SCE assay in human