Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

45392

Chlorphoxim

PESTANAL®, analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C12H14ClN2O3PS
CAS Number:
Molecular Weight:
332.74
EC Number:
238-888-9
UNSPSC Code:
41116107
PubChem Substance ID:
Beilstein/REAXYS Number:
8919975
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

analytical standard

product line

PESTANAL®

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOP(=S)(OCC)ON=C(C#N)c1ccccc1Cl

InChI

1S/C12H14ClN2O3PS/c1-3-16-19(20,17-4-2)18-15-12(9-14)10-7-5-6-8-11(10)13/h5-8H,3-4H2,1-2H3

InChI key

GQKRUMZWUHSLJF-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Dermal

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R C Muirhead-Thomson
Tropenmedizin und Parasitologie, 32(3), 189-193 (1981-09-01)
Tolerance levels Simulium larvae (S. ornatum and S. equinum) to chlorphoxim and decamethrin, relative to those with Abate, were determined in rapid through-flow laboratory test vessels; a 1-hour exposure followed by 24-hour mortality observations being the standard of comparison of
Identity of the OP-insecticide resistant species in the Simulium sanctipauli subcomplex.
R J Post et al.
Annales de la Societe belge de medecine tropicale, 67(1), 71-73 (1987-03-01)
D L Mount et al.
Journal - Association of Official Analytical Chemists, 68(5), 887-890 (1985-09-01)
Liquid chromatographic (LC) methods for determination of active ingredient in chlorphoxim formulations have been developed independently by Bayer AG and the Centers for Disease Control (CDC). Both methods specify separation on a silica gel column. The Bayer method uses a
J Hemingway et al.
Parassitologia, 25(1), 1-8 (1983-04-01)
Adult Anopheles atroparvus, from Cadiz, Spain (strain AT SPA) were resistant to several organophosphorus and carbamate insecticides. Separate lines of the AT SPA strain selected with propoxur, fenitrothion, fenthion and malathion were cross-resistant to these insecticides as well as chlorphoxim
C Pellissier et al.
Toxicological European research. Recherche europeenne en toxicologie, 5(2), 63-69 (1983-03-01)
The Onchocerciasis Control Programme (OCP) is realized in the major part of the treated area by weekly applications of temephos in biotopes of simulles larvae. This insecticide is very effective and its impact on the aquatic fauna is evaluated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service