Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45295

Sigma-Aldrich

Epibromohydrin

purum, ≥97.5% (GC)

Synonym(s):

1-Bromo-2,3-epoxypropane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5BrO
CAS Number:
Molecular Weight:
136.98
Beilstein/REAXYS Number:
79786
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥97.5% (GC)

refractive index

n20/D 1.482 (lit.)
n20/D 1.482

bp

134-136 °C (lit.)

mp

−40 °C (lit.)

density

1.601 g/mL at 25 °C (lit.)

SMILES string

BrCC1CO1

InChI

1S/C3H5BrO/c4-1-3-2-5-3/h3H,1-2H2

InChI key

GKIPXFAANLTWBM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service