44947
Diironnonacarbonyl
purum, ≥97.0% (Fe)
Synonym(s):
Enneacarbonyldiiron, Iron enneacarbonyl, Nonacarbonyldiiron
About This Item
grade
purum
assay
≥97.0% (Fe)
reaction suitability
reagent type: catalyst
core: iron
storage temp.
2-8°C
SMILES string
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].O=C1[Fe]2C(=O)[Fe]1C2=O
InChI
1S/9CO.2Fe/c9*1-2;;
InChI key
JCXLZXJCZPKTBW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Sol. 2 - Water-react 2
Storage Class
4.3 - Hazardous materials which set free flammable gases upon contact with water
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service