Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44784

Supelco

Propyl mercaptan

analytical standard

Synonym(s):

1-Propanethiol, n-Propylmercaptan, Mercaptan C3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2SH
CAS Number:
Molecular Weight:
76.16
Beilstein/REAXYS Number:
1696860
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

vapor density

2.54 (vs air)

vapor pressure

122 mmHg ( 20 °C)

assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.426 (lit.)
n20/D 1.438

bp

67-68 °C (lit.)

mp

−113 °C (lit.)

density

0.820 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

Storage temp.

2-8°C

SMILES string

CCCS

InChI

1S/C3H8S/c1-2-3-4/h4H,2-3H2,1H3

Inchi Key

SUVIGLJNEAMWEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propyl mercaptan, an organic sulfur-containing compound, belongs to the class of volatile odor-active compounds typically found in food and beverage products.[1][2][3][4]

Application

Propyl mercaptan may be used as an analytical standard for the determination of the analyte in water samples,[1] animal wastes,[5] vegetable samples,[2] air as well as organic solvent mixtures,[3] and food waste[4] by various chromatography techniques.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Air pollution from animal wastes. Determination of malodors by gas chromatographic and organoleptic techniques
Burnett WE
Environmental Science & Technology, 3(8), 744-749 (1969)
Odorous compounds in natural waters. Some sulfur compounds associated with blue-green algae
Jenkins D. et al.
Environmental Science & Technology, 1(9), 731-735 (1967)
Concentration and recovery of atmospheric odor pollutants using activated carbon
Brooman DL and Edgerley Jr E
Journal of the Air Pollution Control Association, 16(1), 25-29 (1966)
Characteristic analysis for odor gas emitted from food waste anaerobic fermentation in the pretreatment workshop
Di Y, et al.
Journal of the Air & Waste Management Association (1995), 63(10), 1173-1181 (2013)
Onion flavor and odor, the volatile flavor components of onions.
Carson JF & Wong FF
Journal of Agricultural and Food Chemistry, 9(2), 140-143 (1961)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service