Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

44783

Supelco

Flupirtine

analytical standard

Synonym(s):

Ethyl-N-[2-amino-6-(4-fluorophenylmethylamino) pyridin-3-yl]carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17FN4O2
CAS Number:
Molecular Weight:
304.32
Beilstein/REAXYS Number:
493526
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

Storage temp.

2-8°C

SMILES string

FC1=CC=C(CNC2=NC(N)=C(NC(OCC)=O)C=C2)C=C1

InChI

1S/C15H17FN4O2/c1-2-22-15(21)19-12-7-8-13(20-14(12)17)18-9-10-3-5-11(16)6-4-10/h3-8H,2,9H2,1H3,(H,19,21)(H3,17,18,20)

Inchi Key

JUUFBMODXQKSTD-UHFFFAOYSA-N

General description

Flupirtine belongs to N-methyl-D-aspartate class of drugs.[1] It is a centrally acting non opioid analgesic with muscle relaxant and neuroprotective properties.[2]

Application

Flupirtine may be used as a reference drug for the determination of flupirtine in biological samples by liquid chromatography combined with tandem mass spectrometry (HPLC-MS/MS).[3]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flupirtine Determination in Human Blood Plasma by HPLC with Mass-Spectrometric Detection and its Application to Pharmacokinetic Studies.
Krasnykh L M, et al.
Pharma Chemica, 51(12), 1123-1128 (2018)
Bioanalytical method validation and quantification of flupirtine in canine plasma by HPLC with spectrofluorimetric detection.
De Vito V, et al.
American Journal of Animal and Veterinary Sciences, 10(2), 91-100 (2015)
Antioxidant properties of the triaminopyridine, flupirtine. Biochemical pharmacology
Gassen M, et al.
Biochemical Pharmacology, 56(10), 1323-1329 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service