Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44782

Millipore

E. coli O157:H7 MUG Agar

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200
NACRES:
NA.74

shelf life

limited shelf life, expiry date on the label

Quality Level

composition

agar, 13 g/L
ammonium ferric citrate, 0.5 g/L
bromothymol blue, 0.025 g/L
casein peptone, 20 g/L
meat extract, 2 g/L
4-methylumbelliferyl-β-D-glucuronide, 0.1 g/L
sodium chloride, 5 g/L
sodium deoxycholate, 1.12 g/L
sodium thiosulfate, 2 g/L
sorbitol, 10 g/L
yeast extract, 1 g/L

final pH

7.4±0.2 (25 °C)

application(s)

clinical testing
environmental
food and beverages
food and beverages
food and beverages

microbiology

suitability

selective and differential for Escherichia coli
selective and differential for coliforms
selective and differential for enterovirulent E. coli (EEC)

Application

Selective agar for the isolation and differentiation of enterohaemorrhagic (EHEC) E. coli O157:H7-strains from food and clinical material.

Other Notes

Method to isolate E. coli O157:H7 from food[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

R.A. Szabo et al.
Journal of Food Protection, 10, 768-768 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service