Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44727

Sigma-Aldrich

Chromeon 642-Biotin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

fluorescence

λex 574 nm; λem 676 nm in PBS

suitability

suitable for fluorescence

storage temp.

2-8°C

Legal Information

Licensed from Chromeon GmbH, Regensburg, Germany. Only for R&D use, reselling or repackaging not allowed. Use in any fluorescent lifetime assays requires a separate license.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service