44528
(24aR,28aR)-1,2,9,10,12,13,15,16,23,24,24a,25,26,27,28,28a-Hexadecahydro-3,7:18,22-dimetheno-8,11,14,17,1,24-benzotetraoxadiazacyclohexacosine
About This Item
assay
≥97.0% (HPLC)
composition
carbon content, 70.88% (theory)
hydrogen content, 8.24% (theory)
nitrogen content, 6.36% (theory)
suitability
suitable for NMR (spectroscopy)
SMILES string
C1CC[C@H]2NCc3cccc(OCCOCCOCCOc4cccc(CN[C@@H]2C1)c4)c3
InChI
1S/C26H36N2O4/c1-2-10-26-25(9-1)27-19-21-5-3-7-23(17-21)31-15-13-29-11-12-30-14-16-32-24-8-4-6-22(18-24)20-28-26/h3-8,17-18,25-28H,1-2,9-16,19-20H2/t25-,26-/m1/s1
InChI key
PRVQLIQFJVFWJA-CLJLJLNGSA-N
Packaging
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service