Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44524

Supelco

N,N-Dimethylformamide diethyl acetal

LiChropur, ≥98.0% (GC)

Synonym(s):

1,1-Diethoxy-N,N-dimethylmethylamine, 1,1-Diethoxytrimethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH(OC2H5)2
CAS Number:
Molecular Weight:
147.22
Beilstein/REAXYS Number:
741889
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade ((GC derivatization))

Quality Level

assay

≥98.0% (GC)

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

refractive index

n20/D 1.400 (lit.)
n20/D 1.400

bp

130-133 °C (lit.)

density

0.859 g/mL at 25 °C (lit.)

SMILES string

CCOC(OCC)N(C)C

InChI

1S/C7H17NO2/c1-5-9-7(8(3)4)10-6-2/h7H,5-6H2,1-4H3

InChI key

BWKAYBPLDRWMCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

82.4 °F - closed cup

flash_point_c

28 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service