Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44466

Supelco

Valdecoxib

analytical standard

Synonym(s):

4-(5-Methyl-3-phenyl-4-isoxazolyl)benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14N2O3S
CAS Number:
Molecular Weight:
314.36
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

Cc1onc(-c2ccccc2)c1-c3ccc(cc3)S(N)(=O)=O

InChI

1S/C16H14N2O3S/c1-11-15(12-7-9-14(10-8-12)22(17,19)20)16(18-21-11)13-5-3-2-4-6-13/h2-10H,1H3,(H2,17,19,20)

InChI key

LNPDTQAFDNKSHK-UHFFFAOYSA-N

Gene Information

human ... PTGS2(5743)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Valdecoxib is a non-steroidal anti-inflammatory drug (NSAID), a cyclooxygenase-2 (COX-2) selective inhibitor.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mari-Pau Mena et al.
Experimental cell research, 324(2), 124-136 (2014-03-25)
The mechanisms controlling the switch between the pro-angiogenic and pro-inflammatory states of endothelial cells are still poorly understood. In this paper, we show that: (a) COX-2 expression induced by VEGF-A is NFAT2-dependent; and (b) the integrin profile in endothelial cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service