Recommended Products
form
beads
technique(s)
LPLC: suitable
loss
~80% loss on drying, 110 °C
matrix
styrene-divinylbenzene
matrix active group
sulfonic acid functional group
particle size
50-100 mesh
capacity
1.0 meq/mL by wetted bed volume
separation technique
cation exchange
InChI
1S/C10H11.C10H10.C8H8.Na/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8;/h3,5-8H,1-2,4H2;3-8H,1-2H2;2-7H,1H2;/q-1;;;+1
InChI key
NEQWAZPDMBONCG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service