Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44378

Sigma-Aldrich

M′Fadyean Stain Solution

suitable for microbiology

Synonym(s):

Polychrom Methylene Blue Solution according to M’Fadyean

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
MA.02

product line

BioChemika

form

liquid

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | staining: suitable

application(s)

diagnostic assay manufacturing
hematology
histology

microbiology

storage temp.

room temp

suitability

Bacillus spp.
Corynebacterium spp.

General description

M′Fadyean Stain is used for capsule staining of Bacillus anthracis.[1][2]

Components

Composition:
Methylene blue 3.0 g/l, Ethanol 300 ml/l, Potassium hydroxide 0.1 g/l in water

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P.R. Murray et al.
Manual of Clinical Microbiology, 357-357 (1999)
J. M'Fadyean
Journal of Comparative Pathology, 16, 360-360 (1904)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service