Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

443549

Sigma-Aldrich

Diethyl ether

≥99.0%, anhydrous, ACS reagent, contains 1 ppm BHT as inhibitor

Synonym(s):

Et2O, Ethoxyethane, Ether, Ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Beilstein/REAXYS Number:
1696894
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
grade:
ACS reagent
anhydrous
assay:
≥99.0%
bp:
34.6 °C (lit.)
vapor pressure:
28.69 psi ( 55 °C)
8.59 psi ( 20 °C)

Product Name

Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains 1 ppm BHT as inhibitor

grade

ACS reagent
anhydrous

vapor density

2.6 (vs air)

vapor pressure

28.69 psi ( 55 °C)
8.59 psi ( 20 °C)

assay

≥99.0%

form

liquid

autoignition temp.

320 °F

contains

1 ppm BHT as inhibitor

expl. lim.

36.5 %

dilution

(for analytical testing)

impurities

Alcohol ( as CH3CH2OH ), passes test
≤0.0002 meq/g Titr. acid
≤0.001% Carbonyl ( as HCHO )
≤0.03% water
≤1 ppm Peroxide ( as H2O2 )

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.3530 (lit.)

bp

34.6 °C (lit.)

mp

−116 °C (lit.)

density

0.706 g/mL at 25 °C (lit.)

SMILES string

CCOCC

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

-40.0 °F - closed cup

flash_point_c

-40 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ether-directed ortho-C-H olefination with a palladium(II)/monoprotected amino acid catalyst.
Gang Li et al.
Angewandte Chemie (International ed. in English), 52(4), 1245-1247 (2012-12-15)
Tatsuya Yoshino et al.
Organic letters, 14(16), 4290-4292 (2012-08-09)
A highly efficient total synthesis of the 11-membered cyclic aspercyclides A (1) and B (2) has been achieved by chemo- and regioselective intramolecular oxidative C-O bond formation from differently substituted diphenols.
Faysal Benaskar et al.
ChemSusChem, 6(2), 353-366 (2012-11-30)
A μ(2)-process in the Ullmann-type C-O coupling of potassium phenolate and 4-chloropyridine was successfully performed in a combined microwave (MW) and microflow process. Selective MW absorption in a micro-fixed-bed reactor (μ-FBR) by using a supported Cu nanocatalyst resulted in an
Julia J Griese et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(43), 17189-17194 (2013-10-09)
Although metallocofactors are ubiquitous in enzyme catalysis, how metal binding specificity arises remains poorly understood, especially in the case of metals with similar primary ligand preferences such as manganese and iron. The biochemical selection of manganese over iron presents a
Alessandra Forni et al.
Journal of molecular graphics & modelling, 38, 31-39 (2012-10-23)
The solvent effect on the I⋯O halogen bonding in complexes of iodobenzene derivatives with formaldehyde has been investigated by systematically varying the substituents on the iodobenzene ring. Calculations have been performed at MP2 and DFT levels of theory, using the

Questions

1–7 of 7 Questions  
  1. What are the options for drying solvents?

    1 answer
    1. Sigma-Aldrich offers a full range of high-purity solvents with extremely low water levels specifically manufactured for moisture sensitive Organic and Biotech applications.  Sigma-Aldrich also carries various drying agents such as molecular sieves, which are typically compatible with organic solvents.

      Helpful?

  2. Why do peroxides form in certain solvents and how do I test for it?

    1 answer
    1. A significant number of laboratory solvents can undergo autoxidation under normal storage conditions to form unstable and potentially dangerous peroxide by-products. Molecular structure is the primary factor relating to a material’s potential for hazardous peroxide formation.  It is important to understand the stability of materials in use and if unclear, consult Sigma-Aldrich Technical Support, [email protected]

      Helpful?

  3. Can you recommend a Solvent Extractor for organics from aqueous solution?

    1 answer
    1. Solvent extractors are often used for the quantitative extraction of organics from aqueous solutions and separations in immunoassays. The MIXXOR system has been applied successfully in many laboratory solvent extraction operations and is ideal for the rapid screening of alternative solvents for specific extraction problems.

      Helpful?

  4. What are some of the preservatives used in the most common ethers and chlorinated solvents and why are they added?

    1 answer
    1. Certain solvents will degrade over time requiring special handling and storage considerations. In addition, the products of certain degradation processes pose a potential safety risk if present at sufficiently high levels. For these types of materials, small amounts of stabilizing chemicals are added to slow down or stop material degradation.

      Helpful?

  5. What apparatus do you recommend for HPLC solvent filtration?

    1 answer
    1. The Sigma-Aldrich Vacuum Filtration Assembly gives you all the components you need including: funnel top, fritted glass funnel support, filtration flask, aluminum clamp, and silicone stopper.  The Acrodisc Syringe Filters offer high quality filtration for analytical samples, certified for HPLC to ensure low extractable and available in a broad range of membranes to meet sample compatibility requirements.

      Helpful?

  6. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  7. Who do I contact about larger solvent volume needs?

    1 answer
    1. Email us at [email protected] or visit Sigma-Aldrich Fine Chemicals for development and manufacturing-scale inquiries: [email protected]

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service