Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

443123

Supelco

Ionic strength adjustor buffer solution

for fluoride electrode (1.4% HOAc, 8.2% NaOAc, 5.8% NaCl, trace CDTA)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NB.61

Product Name

Ionic strength adjustor buffer solution for fluoride electrode (1.4% HOAc, 8.2% NaOAc, 5.8% NaCl, trace CDTA),

density

1.2 g/mL at 25 °C

Quality Level

General description

Visit our Sensor Applications portal to learn more.

Application

It may be used as TISAB buffer in determination of fluoride in coal, coal fly ash, phosphate rock, limestone, mineral clay, fossilised materials, oyster tissue and vegetation using pyrohydrolysis for sample decomposition.[1]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morpholinoethanesulfonic acid-based buffer system for improved detection limit and stability of the fluoride ion selective electrode.
Fouskaki, M., et al.
Analytica Chimica Acta, 478.1, 77-84 (2003)
Potentiometric determination of fluoride in geological and biological samples following pyrohydrolytic decomposition.
Dressler, Valderi L., et al.
Analytica Chimica Acta, 466.1, 117-123 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service