About This Item
Recommended Products
grade
analytical standard
Quality Level
vapor density
5.8 (vs air)
vapor pressure
0.2 mmHg ( 20 °C)
assay
≥99.5% (GC)
autoignition temp.
491 °F
shelf life
limited shelf life, expiry date on the label
expl. lim.
~4.7 %
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
refractive index
n20/D 1.429 (lit.)
n20/D 1.430
bp
214-216 °C (lit.)
mp
−35 °C (lit.)
density
0.758 g/mL at 20 °C (lit.)
application(s)
cleaning products
cosmetics
environmental
food and beverages
personal care
petroleum
format
neat
SMILES string
CCCCCCCCCCC=C
InChI
1S/C12H24/c1-3-5-7-9-11-12-10-8-6-4-2/h3H,1,4-12H2,2H3
InChI key
CRSBERNSMYQZNG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Recommended products
signalword
Danger
hcodes
Hazard Classifications
Asp. Tox. 1
supp_hazards
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
174.2 °F - No data available
flash_point_c
79 °C - No data available
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service