Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43964

Supelco

(±)-5-Hydroxyvalproic acid sodium salt

analytical standard

Synonym(s):

5-Hydroxy-2-propylpentanoic acid sodium salt, 5-Hydroxy-2-propylvaleric acid sodium salt, Sodium (±)-5-hydroxyvalproate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NaO3
CAS Number:
Molecular Weight:
182.19
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

O=C(O[Na])C(CCC)CCCO

InChI

1S/C8H16O3/c1-2-4-7(8(10)11)5-3-6-9/h7,9H,2-6H2,1H3,(H,10,11)

InChI key

ZIWBCJXKYKOLJK-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service