Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43957

Supelco

Benzyl acetate

for ion-selective electrodes, Selectophore, ≥99.5%

Synonym(s):

Acetic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2C6H5
CAS Number:
Molecular Weight:
150.17
Beilstein/REAXYS Number:
1908121
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

vapor pressure

23 mmHg ( 110 °C)

Quality Level

product line

Selectophore

assay

≥99.5% (GC)
≥99.5%

form

liquid

autoignition temp.

862 °F

refractive index

n20/D 1.502 (lit.)

bp

206 °C (lit.)

mp

−51 °C (lit.)

density

1.054 g/mL at 25 °C (lit.)

SMILES string

CC(=O)OCc1ccccc1

InChI

1S/C9H10O2/c1-8(10)11-7-9-5-3-2-4-6-9/h2-6H,7H2,1H3

InChI key

QUKGYYKBILRGFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzyl acetate is a colourless liquid having fruity odor. Reaction with oxidants can result in fire and explosion hazards.
Visit our Sensor Applications portal to learn more.

Application

It was used during statistical analysis, through a dose-response curve for odorants, in a study performed to understand the olfactory responses of Drosophila larvae.

Other Notes

Plasticizer for ionophore membranes

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

203.0 °F - closed cup

flash_point_c

95 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T.S.S. Dikshith
Handbook of Chemicals and Safety, 124-125 (2010)
M. Ganjali et al.
Electroanalysis, 16, 1009-1009 (2004)
M. Ganjali et al.
Electroanalysis, 16, 922-922 (2004)
D McGinty et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50 Suppl 2, S363-S384 (2012-03-06)
A toxicologic and dermatologic review of benzyl acetate when used as a fragrance ingredient is presented. Benzyl acetate is a member of the fragrance structural group aryl alkyl alcohol simple acid esters (AAASAE). The AAASAE fragrance ingredients are prepared by
S A Hotchkiss et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 30(2), 145-153 (1992-02-01)
The effect of vehicle and occlusion on the in vitro percutaneous absorption of [methylene-14C]-benzyl acetate (1.7-16.6 mg/cm2) has been studied in diffusion cells using full thickness skin from male Fischer 344 rats. Absorption of neat benzyl acetate through rat skin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service