Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43907

Supelco

Vitexin 4″-O-glucoside

analytical standard

Synonym(s):

4′,5,7-Trihydroxyflavone 8-C-(4″-glucosylglucoside)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30O15
CAS Number:
Molecular Weight:
594.52
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

assay

≥95% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O[C@@H]2CO)c3c(O)cc(O)c4C(=O)C=C(Oc34)c5ccc(O)cc5)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C27H30O15/c28-7-15-19(34)20(35)23(38)27(41-15)42-24-16(8-29)40-26(22(37)21(24)36)18-12(32)5-11(31)17-13(33)6-14(39-25(17)18)9-1-3-10(30)4-2-9/h1-6,15-16,19-24,26-32,34-38H,7-8H2/t15-,16-,19-,20+,21-,22-,23-,24-,26+,27+/m1/s1

InChI key

NDSUKTASTPEKBX-LXXMDOISSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service