Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

436666

Supelco

Dowex Marathon WBA free base

free base

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

beads

moisture

50-60%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (macroporous)

matrix active group

polyamine functional group

particle size distribution

26-32 mesh

particle size

475-575 μm

operating pH

0-7

capacity

1.5 meq/mL by wetted bed volume

separation technique

anion exchange

InChI

1S/C10H12.C10H10.C8H8.C2H7N/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8;1-3-2/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2;3H,1-2H3

InChI key

RQUJHSIERAWNRD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Weakly basic anion exchange resin that is well suited for combined use with strong base anion resins for water with high concentration of mineral anions or high organic fouling potential. It is excellent for all demineralization applications, and meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Features and Benefits

Weakly basic anion, macroporous

Other Notes

Monodispersed beads make this resin superior and more efficient than the Dowex MWA-1.
This ion-exchange resin has not been specially processed nor cleaned. We suggest that it be treated to a suitable preliminary elution and wash.

Legal Information

Marathon is a trademark of The Dow Chemical Company or an affiliated company of Dow

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service