43506
Polystyrene Beads for Gel Permeation Chromatography
for organic compounds with molecular weight up to 2000 daltons
Synonym(s):
Gel Permeation Chromatography Polystyrene Beads, Size exclusion Chromatography Polystyrene Beads
About This Item
Recommended Products
Quality Level
form
beads
technique(s)
LPLC: suitable
matrix
crosslinked with 3% DVB
matrix active group
polymer
particle size
40-80 μm
separation technique
size exclusion (SEC)
General description
Application
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service