Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43359

Supelco

Manganese(II) ionophore II

Selectophore, function tested

Synonym(s):

2,2′-Bis(salicylideneamino)azobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H20N4O2
CAS Number:
Molecular Weight:
420.46
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

form

solid

quality

function tested

SMILES string

Oc1ccccc1\C=N\c2ccccc2\N=N\c3ccccc3\N=C\c4ccccc4O

InChI

1S/C26H20N4O2/c31-25-15-7-1-9-19(25)17-27-21-11-3-5-13-23(21)29-30-24-14-6-4-12-22(24)28-18-20-10-2-8-16-26(20)32/h1-18,31-32H/b27-17+,28-18+,30-29+

Inchi Key

SOZKMMFWBXRDSE-VBJJXLHFSA-N

General description

Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina Minnelli et al.
International journal of pharmaceutics, 552(1-2), 225-234 (2018-10-07)
Epigallocatechin-3-gallate (EGCG) is a polyphenolic catechin from green tea, well known for being bioactive in age-associated pathologies where oxidative stress plays a preeminent role. The activity of this molecule is however contrasted by its high chemical and metabolic instability that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service