Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

42994

Supelco

Disperse Orange 11

analytical standard

Synonym(s):

1-Amino-2-methylanthraquinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11NO2
CAS Number:
Molecular Weight:
237.25
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥96.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

204-206 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Cc1ccc2C(=O)c3ccccc3C(=O)c2c1N

InChI

1S/C15H11NO2/c1-8-6-7-11-12(13(8)16)15(18)10-5-3-2-4-9(10)14(11)17/h2-7H,16H2,1H3

InChI key

ZLCUIOWQYBYEBG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Carc. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1-Amino-2-methylanthraquinone.
Report on carcinogens : carcinogen profiles, 12, 41-41 (2011-08-11)
1-Amino-2-methylanthraquinone.
Report on carcinogens : carcinogen profiles, 11, III16-III16 (2004-01-01)
1-Amino-2-methylanthraquinone.
Report on carcinogens : carcinogen profiles, 10, 14-14 (2004-08-20)
The acute toxicity of 1-methylaminoanthraquinone in dogs and rabbits and its metabolism in sheep.
B W Martin et al.
Archives of environmental contamination and toxicology, 12(4), 499-507 (1983-07-01)
1-Amino-2-methylanthraquinone.
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans, 27, 199-204 (1982-04-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service