Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42986

Supelco

Methyl cis,cis-11,12;14,15-diepoxyeicosanoate

qualitative standard, ≥97.0% (sum of stereoisomers, GC)

Synonym(s):

cis,cis-11,14-Eicosadienoic acid diepoxide methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H38O4
Molecular Weight:
354.52
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

Quality Level

assay

≥97.0% (sum of stereoisomers, GC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

−20°C

SMILES string

[H][C@@]2(C[C@@H]1O[C@H]1CCCCCCCCCC(=O)OC)O[C@@H]2CCCCC

InChI

1S/C21H38O4/c1-3-4-10-13-17-19(24-17)16-20-18(25-20)14-11-8-6-5-7-9-12-15-21(22)23-2/h17-20H,3-16H2,1-2H3/t17-,18+,19+,20?/m1/s1

InChI key

RYSWSVUNOVFICZ-QYRWZOIFSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Internal standard for the assay of epoxidized soy bean oil (ESBO) in foods with HPLC-MS or GC-MS. ESBO is currently still employed as plasticizer and HCl scavenger in foamed PVC used in cap linings

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katherine S Carlos et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 35(6), 1214-1222 (2018-03-07)
PVC is a common food contact material that is usually plasticised to increase its flexibility. Phthalates are one class of chemical compounds that are often used as plasticisers in PVC in a wide range of industries. They may be used

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service