Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42942

Supelco

Iron(III) ionophore VI

Selectophore, function tested

Synonym(s):

1-(4-Dimethylaminophenyl)-2-(6-phenanthridinyl)ethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H20N2O
CAS Number:
Molecular Weight:
340.42
Beilstein/REAXYS Number:
10464593
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

quality

function tested

storage temp.

2-8°C

SMILES string

CN(C)c1ccc(cc1)C(=O)Cc2nc3ccccc3c4ccccc24

InChI

1S/C23H20N2O/c1-25(2)17-13-11-16(12-14-17)23(26)15-22-20-9-4-3-7-18(20)19-8-5-6-10-21(19)24-22/h3-14H,15H2,1-2H3

InChI key

YXUDOLSUGNSQGS-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service