Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42925

Supelco

Schaftoside

analytical standard

Synonym(s):

8-α-L-Arabinopyranosyl-6-β-D-glucopyranosyl-5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, Apigenin 8-C-α-L-arabinoside 6-C-β-D-glucoside, Shaftoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H28O14
CAS Number:
Molecular Weight:
564.49
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)c([C@@H]3OC[C@H](O)[C@H](O)[C@H]3O)c4OC(=CC(=O)c4c2O)c5ccc(O)cc5

InChI

1S/C26H28O14/c27-6-13-18(32)21(35)23(37)26(40-13)15-19(33)14-10(29)5-12(8-1-3-9(28)4-2-8)39-24(14)16(20(15)34)25-22(36)17(31)11(30)7-38-25/h1-5,11,13,17-18,21-23,25-28,30-37H,6-7H2/t11-,13+,17-,18+,21-,22+,23+,25-,26-/m0/s1

InChI key

MMDUKUSNQNWVET-VYUBKLCTSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Schaftoside may be used as a reference standard for the determination of the analyte in quince seeds using ion trap high-performance liquid chromatography with pulsed amperometric detection and electrospray ionization combined with tandem mass spectrometry (HPLC-PAD-ESI/MS/MS).[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Approach to the study of C?glycosyl flavones by ion trap HPLC?PAD?ESI/MS/MS: application to seeds of quince (Cydonia oblonga).
Ferreres F, et al.
Phytochemical Analysis, 14(6), 352-359 (2003)

Protocols

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service