Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42904

Supelco

Oxypeucedanin

analytical standard

Synonym(s):

4-[[(2S)-3,3-Dimethyl-2-oxiranyl]methoxy]-7H-furo[3,2-g][1]benzopyran-7-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14O5
CAS Number:
Molecular Weight:
286.28
UNSPSC Code:
85151701

grade

analytical standard

assay

≥95.0% (HPLC)

optical activity

[α]/D -16.0±2.0°, c = 0.14 in chloroform

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

storage temp.

−20°C

InChI

1S/C16H14O5/c1-16(2)13(21-16)8-19-15-9-3-4-14(17)20-12(9)7-11-10(15)5-6-18-11/h3-7,13H,8H2,1-2H3

InChI key

QTAGQHZOLRFCBU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service