Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42883

Supelco

Delphinidin 3-sambubioside chloride

analytical standard

Synonym(s):

Delphinidin 3-O-(2-O-β-D-xylopyranosyl)-β-D-glucopyranoside chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H29O16Cl
CAS Number:
Molecular Weight:
632.95
UNSPSC Code:
12352200
NACRES:
NA.24
Product 42883 is not currently sold in your country. Contact Technical Service

grade

analytical standard

assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC1=CC(O)=C(C=C(O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O[C@@H]3OC[C@@H](O)[C@H](O)[C@H]3O)C(C4=CC(O)=C(O)C(O)=C4)=[O+]5)C5=C1.[Cl-]

Looking for similar products? Visit Product Comparison Guide

General description

Delphinidin 3-sambubioside is an anthocyanin flavonoid, which is naturally present as a pigment in fruits, flowers and leaves. This water-soluble polyphenolic compound is known to exert powerful antioxidant performance.[1][2]

Application

Delphinidin 3-sambubioside chloride may be used as an analytical standard for the determination of the analyte in calyces of Hibiscus sabdariffa flowers by high performance liquid chromatography (HPLC) technique.[3]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of anthocyanins and proanthocyanidins in some cultivars of Ribes, Aronia, and Sambucus and their antioxidant capacity
Wu X, et al.
Journal of Agricultural and Food Chemistry, 52(26), 7846-7856 (2004)
Impact of texturing using instant pressure drop treatment prior to solvent extraction of anthocyanins from Malaysian Roselle (Hibiscus sabdariffa)
Amor BB and Allaf K
Food Chemistry, 115(3), 820-825 (2009)
Preparation and validated analysis of anthocyanin concentrate from the calyces of Hibiscus sabdariffa
Opletal L, et al.
Natural Product Communications, 12(1), 1934578X1701200112-1934578X1701200112 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service