Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42725

Supelco

N-Desmethyl-4-hydroxytamoxifen, mixture of isomers

analytical standard

Synonym(s):

4-{{4-[2-(Methylamino)ethoxy]phenyl}-2-phenyl-1-buten-1-yl}phenol, Desmethylhydroxytamoxifen, Endoxifen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H27NO2
CAS Number:
Molecular Weight:
373.49
Beilstein/REAXYS Number:
9655609
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥95.0% (sum of isomners, HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤3.0% water

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

CCC(C1=CC=CC=C1)=C(C2=CC=C(O)C=C2)C3=CC=C(OCCNC)C=C3

InChI

1S/C25H27NO2/c1-3-24(19-7-5-4-6-8-19)25(20-9-13-22(27)14-10-20)21-11-15-23(16-12-21)28-18-17-26-2/h4-16,26-27H,3,17-18H2,1-2H3

InChI key

MHJBZVSGOZTKRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 4 - Repr. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

E Giannoutsou et al.
Protoplasma, 257(1), 141-156 (2019-09-01)
This article deals with the distribution of callose and of the homogalacturonan (HG) epitopes recognized by LM20, JIM5, and 2F4 antibodies in cell walls of differentiating and functioning stomatal complexes of the monocotyledon Zea mays and the dicotyledon Vigna sinensis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service