Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

42713

Supelco

1-Deoxy-D-xylulose-5-phosphate sodium salt

analytical standard

Synonym(s):

DXP sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11O7P · xNa+
CAS Number:
Molecular Weight:
214.11 (free acid basis)
Beilstein/REAXYS Number:
8367371
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (TLC)

optical activity

[α]/D +38.0±4°, c = 0.1 in 0.1 M HCl (after 3 hours)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

InChI

1S/C5H11O7P/c1-3(6)5(8)4(7)2-12-13(9,10)11/h4-5,7-8H,2H2,1H3,(H2,9,10,11)/t4-,5-/m1/s1

InChI key

AJPADPZSRRUGHI-RFZPGFLSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Metabolite of the non-mevalonate pathway, generally found in prokaryotes, as precursor to isoprenoids as well asnon-isoprenoids like vitamins. As this pathway is not present in humans, it is of interest for the development of bacterium-specific drugs in the search for treatments of infectious diseases.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service