Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

426326

Sigma-Aldrich

Tetrabutylammonium hydroxide solution

ACS reagent, 1.0 M in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(OH)
CAS Number:
Molecular Weight:
259.47
Beilstein/REAXYS Number:
3571228
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

ACS reagent

Quality Level

form

liquid

concentration

1.0 M in H2O
1.0 M±0.02 M (ACS specification)

impurities

≤0.2% tributylamine

color

APHA: ≤30

refractive index

n20/D 1.389

density

1 g/mL at 25 °C

anion traces

carbonate (CO32-): ≤0.1%
chloride (Cl-): ≤0.08%

cation traces

K: ≤2 ppm
Na: ≤2 ppm

SMILES string

[OH-].CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.H2O/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H2/q+1;/p-1

InChI key

VDZOOKBUILJEDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetrabutylammonium hydroxide (TBAH) solution it is a strong organic base.[1] It enables the dispersion of fatty acids in water at room temperature which allows the formation of emulsions and foams.[2] TBAH is used as a titrant in the potentiometric titration of weak acids in non-aqueous media.[3]

Application

Phase-transfer catalyst.[4]
Tetrabutylammonium hydroxide solution may be used in the following processes:
  • Synthesis of carbon doped TiO2 (titanium dioxide).[5]
  • Synthesis of C60 derivative-hydroxylated fullerene epoxide.[6]
  • As a hydrolysis controlling agent in the prearation of TiO2 sol.[7]

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surface doping of conjugated polymers by graphene oxide and its application for organic electronic devices.
Gao Y, et al.
Advanced Mat., 23(16), 1903-1908 (2011)
Synthesis and characterization of hydroxylated fullerene epoxide-an intermediate for forming fullerol.
Li T, et al.
Journal of Central South University of Technology, 6(1), 35-36 (1999)
Low temperature deposition of TiO2 thin films on polyvinyl alcohol fibers with photocatalytical and antibacterial activities.
Liuxue Z, et al.
Applied Surface Science, 254(6), 1771-1774 (2008)
Journal of the Chemical Society. Chemical Communications, 1045-1045 (1990)
Daylight photocatalysis by carbon-modified titanium dioxide.
Sakthivel S and Kisch H.
Angewandte Chemie (International Edition in English), 42(40), 4908-4911 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service