Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42624

Supelco

Diisobutyl adipate

analytical standard

Synonym(s):

Adipic acid bis(2-methylpropyl) ester, Adipic acid diisobutyl ester, Hexanedioic acid 1,6-bis(2-methylpropyl) ester, NSC 6343

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CH2CO2CH2CH(CH3)2]2
CAS Number:
Molecular Weight:
258.35
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.432 (lit.)

bp

293 °C (lit.)

density

0.954 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CC(C)COC(=O)CCCCC(=O)OCC(C)C

InChI

1S/C14H26O4/c1-11(2)9-17-13(15)7-5-6-8-14(16)18-10-12(3)4/h11-12H,5-10H2,1-4H3

InChI key

RDOFJDLLWVCMRU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diisobutyl adipate (DIBA), a bis(2-methylpropyl) ester of adipic acid, is a plasticizer. It finds use as an additive in plastics and foodstuffs on account of its attractive features such as stability, fluidity and low volatility.[1][2][3]

Application

Diisobutyl adipate may be used as an analytical reference standard for the determination of the analyte in poly(vinyl chloride) plastics, human serum and food samples by gas chromatography (GC) based analytical techniques.[4][2][1]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of phthalates and adipates in human serum using gas chromatography-mass spectrometry with solid-phase extraction
Guo ZY, et al.
Biomedical Chromatography, 24(10), 1094-1099 (2010)
Determination of plasticisers in food by gas chromatography-mass spectrometry with ion-trap mass detection
Lau OW and Wong SK
Journal of Chromatography A, 737(2), 338-342 (1996)
Synthetic ester-based oils and their application in power industry
Mentlik V, et al.
International Conference on Renewable Energies and Power Quality (ICREPQ?09) (2009)
Determination of adipate plasticizers in poly (vinyl chloride) by microwave-assisted extraction
Cano JM, et al.
Journal of Chromatography A, 963(1-2), 401-409 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service