Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42243

Supelco

9-Methyl-1,2,3,9-tetrahydro-4H-carbazol-4-one

pharmaceutical impurity standard

Synonym(s):

Ondansetron Related Compound C (USP), Ondansetron impurity C (PhEur), 1,2,3,9-Tetrahydro-9-methyl-4H-carbazol-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13NO
CAS Number:
Molecular Weight:
199.25
Beilstein/REAXYS Number:
153862
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10% related substances HPLC impurities

loss

≤5% loss on drying

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cn1c2CCC(=C)C(=O)c2c3ccccc13

InChI

1S/C14H13NO/c1-9-7-8-12-13(14(9)16)10-5-3-4-6-11(10)15(12)2/h3-6H,1,7-8H2,2H3

Inchi Key

AGQJDIDJKSFVTC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service