Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42166

Supelco

Mass scale and resolution set-up solution

suitable for mass spectrometry (MS), 2.0 g/L NaI and 0.05 g/L RbI in 50:50 2-propanol and water

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105

form

liquid

Quality Level

quality

2.0 g/L NaI and 0.05 g/L RbI in 50:50 2-propanol and water

technique(s)

LC/MS: suitable
mass spectrometry (MS): suitable

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

53.6 °F

flash_point_c

12 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service