Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

42127

Supelco

Oil Sand Process-Affected Water Calibration Standard Kit

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material

packaging

ampule of 2 mL

storage temp.

2-8°C

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC2353 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Analyte

Description

    Toluene 2000 µg/mL
    o-Xylene 2000 µg/mL
    2-Methynaphthalene 2000 µg/mL
    2,3-Dimethylnaphthalene 2000 µg/mL
    9-Methylanthracene 2000 µg/mL

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service