Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41760

Supelco

Maize GMO Standard ERM-BF424

Set for 0%, 0.1%, 1%, 10% 59122 Maize

Synonym(s):

GMO Standard ERM-BF424

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
Product 41760 is not currently sold in your country. Contact Technical Service

grade

certified reference material

application(s)

cleaning products
cosmetics
food and beverages
microbiology
personal care
pharmaceutical (small molecule)

format

matrix material

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service