Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41736

Supelco

Diethyl sulfide

analytical standard

Synonym(s):

1,1′-Thiobisethane, Ethyl sulfide, NSC 75157

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2S
CAS Number:
Molecular Weight:
90.19
Beilstein/REAXYS Number:
1696909
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

105 mmHg ( 37.7 °C)

assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.442 (lit.)

bp

90-92 °C (lit.)

mp

−100 °C (lit.)

density

0.837 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CCSCC

InChI

1S/C4H10S/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

LJSQFQKUNVCTIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl sulfide, a volatile alkyl sulfide, is an aroma active compound in a variety of foodstuffs and beverages. This sulfur-compound can significantly affect the quality of food and drinks by virtue of its low sensory threshold.[1][2][3][4]

application

Diethyl sulfide may be used as an analytical standard for the determination of the analyte in wastewater,[1] wines,[2][3][5] and beer[6][4] by gas chromatography (GC) based techniques.

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

15.8 °F - closed cup

flash_point_c

-9 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Chromatographic analysis of volatile sulphur compounds in wines using the static headspace technique with flame photometric detection
Mestres M, et al.
Journal of Chromatography A, 773(1-2), 261-269 (1997)
Rapid quantification of flavor-active sulfur compounds in beer
Dercksen AW, et al.
Journal of the American Society of Brewing Chemists, 50(3), 93-101 (1992)
Selective determination of volatile sulfur compounds in wine by gas chromatography with sulfur chemiluminescence detection
Siebert TE, et al.
Journal of Agricultural and Food Chemistry, 58(17), 9454-9462 (2010)
Determination of volatile alkyl sulfides in wastewater by headspace solid-phase microextraction followed by gas chromatography-mass spectrometry
Abalos M, et al.
Journal of Chromatography A, 963(1-2), 249-257 (2002)
Headspace solid-phase microextraction analysis of volatile sulphides and disulphides in wine aroma
Mestres M, et al.
Journal of Chromatography A, 808(1-2), 211-218 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service