Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41716

Supelco

N,N-Dimethyltrimethylsilylamine

Selectophore, for ion-selective electrodes

Synonym(s):

(Dimethylamino)trimethylsilane, N-(Trimethylsilyl)dimethylamine, TMSDMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiN(CH3)2
CAS Number:
Molecular Weight:
117.26
Beilstein/REAXYS Number:
1731861
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

refractive index

n20/D 1.397 (lit.)

bp

84 °C (lit.)

density

0.732 g/mL at 25 °C (lit.)

SMILES string

CN(C)[Si](C)(C)C

InChI

1S/C5H15NSi/c1-6(2)7(3,4)5/h1-5H3

InChI key

KAHVZNKZQFSBFW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameCorrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - Water-react 1

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

10.4 °F - closed cup

flash_point_c

-12 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James P. Landers
Handbook of Capillary Electrophoresis, 428-428 (1996)
Brian Roland Larsen et al.
Glia, 62(4), 608-622 (2014-02-01)
Network activity in the brain is associated with a transient increase in extracellular K(+) concentration. The excess K(+) is removed from the extracellular space by mechanisms proposed to involve Kir4.1-mediated spatial buffering, the Na(+)/K(+)/2Cl(-) cotransporter 1 (NKCC1), and/or Na(+)/K(+)-ATPase activity.
Zhuo Luan et al.
Experimental neurology, 261, 791-801 (2014-09-11)
The activity of Na(+)/K(+)-ATPase establishes transmembrane ion gradients and is essential to cell function and survival. Either dysregulation or deficiency of neuronal Na(+)/K(+)-ATPase has been implicated in the pathogenesis of many neurodegenerative disorders such as Alzheimer's disease, Parkinson's disease and
Brian Roland Larsen et al.
The Journal of physiology, 597(2), 583-597 (2018-10-26)
Neuronal activity induces fluctuation in extracellular space volume, [K+ ]o and pHo , the management of which influences neuronal function The neighbour astrocytes buffer the K+ and pH and swell during the process, causing shrinkage of the extracellular space In
Trine Lisberg Toft-Bertelsen et al.
Glia, 69(1), 28-41 (2020-06-09)
The mammalian brain consists of 80% water, which is continuously shifted between different compartments and cellular structures by mechanisms that are, to a large extent, unresolved. Aquaporin 4 (AQP4) is abundantly expressed in glia and ependymal cells of the mammalian

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service