Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41608

Supelco

Ethylene glycol diacrylate

analytical standard

Synonym(s):

Ethylene diacrylate, Ethylene glycol dipropenoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2C=CHCO2CH2-)2
CAS Number:
Molecular Weight:
170.16
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.453 (lit.)

bp

66-68 °C/2 mmHg (lit.)

density

1.094 g/mL at 25 °C (lit.)

application(s)

environmental
pharmaceutical

format

neat

SMILES string

C=CC(=O)OCCOC(=O)C=C

InChI

1S/C8H10O4/c1-3-7(9)11-5-6-12-8(10)4-2/h3-4H,1-2,5-6H2

InChI key

KUDUQBURMYMBIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

The purity is calculated based on the principle of mass balance and is mentioned on the CoA

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

213.8 °F - closed cup

flash_point_c

101 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yandong Zhang et al.
Communications biology, 3(1), 420-420 (2020-08-05)
While technologies for measuring transcriptomes in single cells have matured, methods for measuring proteins and their post-translational modification (PTM) states in single cells are still being actively developed. Unlike nucleic acids, proteins cannot be amplified, making detection of minute quantities

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service