Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41601

Supelco

2,2,4-Trimethyl-1,3-pentanediol diisobutyrate

analytical standard

Synonym(s):

1-Isopropyl-2,2-dimethyltrimethylene diisobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH[O2CCH(CH3)2]C(CH3)2CH2O2CCH(CH3)2
CAS Number:
Molecular Weight:
286.41
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.5% (GC)

autoignition temp.

795 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

0.48 %, 172 °F
3.1 %, 230 °F

refractive index

n20/D 1.433-1.435
n20/D 1.434 (lit.)

bp

280 °C (lit.)

mp

−70 °C (lit.)

density

0.941 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CC(C)C(OC(=O)C(C)C)C(C)(C)COC(=O)C(C)C

InChI

1S/C16H30O4/c1-10(2)13(20-15(18)12(5)6)16(7,8)9-19-14(17)11(3)4/h10-13H,9H2,1-8H3

InChI key

OMVSWZDEEGIJJI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2,4-Trimethyl-1,3-pentanediol monoisobutyrate (TMPD-MIB), a volatile organic compound (VOC), is an important component found in water-based paints, printing inks, and anti-freeze agents. TMPD-MIB is a non-phthalate alternate plasticizer used in polyvinyl chloride plastics. Its detection in polystyrene and polypropylene cups used for food packing has been investigated.[1][2][3]
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

2,2,4-Trimethyl-1,3-pentanediol monoisobutyrate may be used as an analytical reference standard for the determination of the analyte in water-based paints,[1] Morchella mushroom,[4] ground-water samples,[5] and plastic baby bottles[6] by chromatography techniques.

pictograms

Health hazard

signalword

Warning

Hazard Classifications

Aquatic Chronic 3 - Repr. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

276.8 °F - Non-equilibrium method

flash_point_c

136 °C - Non-equilibrium method


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic contaminants in the groundwaters of a lignite-bearing basin from northern Greece
Iordanidis A, et al.
Desalination and Water Treatment, 57(12), 5435-5443 (2016)
Detection of volatile aroma compounds of Morchella by headspace gas chromatography mass spectrometry (HS-GC/MS)
Taskin, Hatira
Notulae Botanicae Horti Agrobotanici Cluj-Napoca , 41(1), 122-122 (2013)
Structural characterization of potential endocrine disrupting activity of alternate plasticizers di-(2-ethylhexyl) adipate (DEHA), acetyl tributyl citrate (ATBC) and 2, 2, 4-trimethyl 1, 3-pentanediol diisobutyrate (TPIB) with human sex hormone-binding globulin
Sheikh IA and Beg MA
Reproductive Toxicology, 83(3), 46-53 (2019)
Quantitative determination of migrating compounds from plastic baby bottles by validated GC-QqQ-MS and LC-QqQ-MS methods
Onghena M, et al.
Food Analytical Methods, 9(9), 2600-2612 (2016)
Volatile organic compound (VOC) analysis and anti-VOC measures in water-based paints
Nakashima H, et al.
Journal of Health Science, 53(3), 311-319 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service