Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41594

Sigma-Aldrich

Methyl 5-hexenoate

≥95.0% (GC)

Synonym(s):

5-Hexenoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2:CH(CH2)3COOCH3
CAS Number:
Molecular Weight:
128.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥95.0% (GC)

refractive index

n20/D 1.421

functional group

allyl
ester

SMILES string

COC(=O)CCCC=C

InChI

1S/C7H12O2/c1-3-4-5-6-7(8)9-2/h3H,1,4-6H2,2H3

InChI key

ASKDFGVMJZMYEM-UHFFFAOYSA-N

pictograms

FlameExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

115.2 °F

flash_point_c

46.2 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Storage and ultraviolet-induced tissue stress effects on fresh-cut pineapple.
Lamikanra O and Richard OA.
Journal of the Science of Food and Agriculture, 84(14), 1812-1816 (2004)
Detailed chemical kinetic oxidation mechanism for a biodiesel surrogate.
Herbinet O, et al.
Combustion and Flame, 154(3), 507-528 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service