Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41561

Supelco

Atorvastatin-(anilide ring-d5) calcium salt

analytical standard

Synonym(s):

[2H5]-Atorvastatin calcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33D5H29FN2O5 · 0.5 Ca
CAS Number:
Molecular Weight:
582.70
UNSPSC Code:
41116107

grade

analytical standard

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

[Ca++].[2H]c1c([2H])c([2H])c(NC(=O)c2c(C(C)C)n(CC[C@@H](O)C[C@@H](O)CC([O-])=O)c(-c3ccc(F)cc3)c2-c4ccccc4)c([2H])c1[2H].[2H]c5c([2H])c([2H])c(NC(=O)c6c(C(C)C)n(CC[C@@H](O)C[C@@H](O)CC([O-])=O)c(-c7ccc(F)cc7)c6-c8ccccc8)c([2H])c5[2H]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

LC-MS: identity conforms

pictograms

Health hazard

signalword

Danger

Hazard Classifications

Repr. 2 - STOT RE 1 Oral

target_organs

Gastro-intestinal system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service