Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41392

Supelco

4-Bromophenacyl trifluoromethanesulfonate

for HPLC derivatization, LiChropur, ≥95% (H-NMR/C-NMR)

Synonym(s):

4-Bromophenacyl triflate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6BrF3O4S
CAS Number:
Molecular Weight:
347.11
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade ((HPLC))
for HPLC derivatization

Quality Level

assay

≥95% (H-NMR/C-NMR)

form

solid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

SMILES string

FC(F)(F)S(=O)(=O)OCC(=O)c1ccc(Br)cc1

InChI

1S/C9H6BrF3O4S/c10-7-3-1-6(2-4-7)8(14)5-17-18(15,16)9(11,12)13/h1-4H,5H2

Inchi Key

YMYKWFCLILHEJB-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Assay of free and ′total′ carnitine after derivatization with 4′-bromophenacyl triflate and HPLC[1]; Detn. of carnitine capillary electrophoresis[2]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of free carnitine and 'total' carnitine in human urine: derivatization with 4'-bromophenacyl trifluoromethanesulfonate and high performance liquid chromatography.
P E Minkler et al.
Clinica chimica acta; international journal of clinical chemistry, 212(1-2), 55-64 (1992-11-16)
P E Minkler et al.
Analytical biochemistry, 231(2), 315-322 (1995-11-01)
A method for the quantitative determination of carnitine, acetylcarnitine, and total carnitine in tissue was developed for application to clinical research and diagnosis. Human skeletal muscle and heart specimens (10-20 mg) were homogenized in 1 ml of water. Aliquots of
L Vernez et al.
Journal of chromatography. A, 895(1-2), 309-316 (2000-12-06)
A capillary electrophoresis method is described for the simultaneous analysis of carnitine and short-chain acylcarnitines in aqueous standard solutions and urine samples. Samples were worked up using silica gel extraction and derivatization with 4'-bromophenacyl trifluoromethanesulfonate. Separation was performed in less
S Krahenbuhl et al.
Journal of chromatography, 573(1), 3-10 (1992-01-03)
A method for the isolation and chromatography of butyrobetaine from plasma, urine, and liver is described. The recovery of [3H-methyl]butyrobetaine from spiked biological samples was from 76-80%. Spiked samples then were derivatized with 4'-bromophenacyl trifluoromethanesulfonate and the butyrobetaine 4'-bromophenacyl ester
Paul E Minkler et al.
Analytical chemistry, 77(5), 1448-1457 (2005-03-01)
A strategy for detection of carnitine and acylcarnitines is introduced. This versatile system has four components: (1) isolation by protein precipitation/desalting and cation-exchange solid-phase extraction, (2) derivatization of carnitine and acylcarnitines with pentafluorophenacyl trifluoromethanesulfonate, (3) sequential ion-exchange/reversed-phase chromatography using a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service