Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41152

Supelco

2-Hydroxypyridine

analytical standard

Synonym(s):

2(1H)-Pyridone, 2-Pyridinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5NO
CAS Number:
Molecular Weight:
95.10
Beilstein/REAXYS Number:
105757
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

bp

280-281 °C (lit.)

mp

105-107 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Oc1ccccn1

InChI

1S/C5H5NO/c7-5-3-1-2-4-6-5/h1-4H,(H,6,7)

InChI key

UBQKCCHYAOITMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Hydroxypyridine, an enolic tautomer of 2-pyridone, belongs to the class of volatile organic compounds (VOCs).[1][2]

Application

2-Hydroxypyridine may be used as an analytical standard for the determination of the analyte in roasted coffee beans[1] and Agave tequilana Weber var. azul − the raw material for the generation of tequila beverage[3] by gas chromatography-mass spectrometry (GC-MS) technique.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile compounds as potential defective coffee beans? markers
Toci AT and FA
Food Chemistry, 108(3), 1133-1141 (2008)
Photoionization of 2-pyridone and 2-hydroxypyridine
Poully JC, et al.
Physical Chemistry Chemical Physics, 12(14), 3566-3572 (2010)
Generation of Maillard compounds from inulin during the thermal processing of Agave tequilana Weber Var. azul.
Mancilla-Margalli NA & Lopez MG
Journal of Agricultural and Food Chemistry, 50(4), 806-812 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service