Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40926

Millipore

Fecal Coliforms Enzyme Confirmation Test

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NA.85

shelf life

limited shelf life, expiry date on the label

Quality Level

application(s)

food and beverages
microbiology

suitability

Escherichia coli
coliforms

Application

This tests utilizes a fluorogenic substrate which, when hydrolyzed by a specific enzyme (during peptide hydrolysis), produces a blue/white fluorescence.

Components

1 Bottle of Reagent A (Buffer)
12 Test Cards with 4 spots

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service