Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40781

Supelco

4-Methylanisole

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4OCH3
CAS Number:
Molecular Weight:
122.16
Beilstein/REAXYS Number:
1237336
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

5.25 mmHg ( 50 °C)

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.510-1.514
n20/D 1.511 (lit.)

bp

174 °C (lit.)

density

0.969 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

COc1ccc(C)cc1

InChI

1S/C8H10O/c1-7-3-5-8(9-2)6-4-7/h3-6H,1-2H3

InChI key

CHLICZRVGGXEOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Methylanisole is an odorant benzenoid belonging to the group of volatile organic compounds (VOCs). It can contribute to the floral scent of the plants. 4-Methylanisole is typically a lignin-derived bio-oil that is also a major ingredient of certain food products.

Application

4-Methylanisole may be used as an analytical standard for the determination of the analyte in Malus taxa flowers, coffee brews, aqueous matrices, food products, and Ficus semicordata by various chromatography techniques.

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

138.2 °F - closed cup

flash_point_c

59 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discrimination of Malus taxa with different scent intensities using electronic nose and gas chromatography-mass spectrometry
Fan J, et al.
Sensors, 18(10), 3429-3429 (2018)
Changes in headspace volatile concentrations of coffee brews caused by the roasting process and the brewing procedure
Lopez-Galilea I, et al.
Journal of Agricultural and Food Chemistry, 54(22), 8560-8566 (2006)
Oxidation of the odorous compound 2, 4, 6-trichloroanisole by UV activated persulfate: kinetics, products, and pathways
Luo C, et al.
Water Research, 96(22), 12-21 (2016)
Catalytic conversion of compounds representative of lignin-derived bio-oils: a reaction network for guaiacol, anisole, 4-methylanisole, and cyclohexanone conversion catalysed by Pt/gamma-Al2O3
Runnebaum RC, et al.
Catalysis Science & Technology, 2(1), 113-118 (2012)
Private channel: a single unusual compound assures specific pollinator attraction in Ficus semicordata
Chen C, et al.
Functional Ecology, 23(5), 941-950 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service