Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40764

Supelco

(+)-Tröger′s base

LiChropur, ≥99.0%

Synonym(s):

(5S,11S)-(+)-2,8-Dimethyl-6H,12H-5,11-methanodibenzo[b,f][1,5]diazocine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18N2
CAS Number:
Molecular Weight:
250.34
Beilstein/REAXYS Number:
88612
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade ((chiral))

Quality Level

assay

≥99.0% (sum of enantiomers, HPLC)
≥99.0%

optical activity

[α]20/D +283±4°, c = 0.3% in hexane

optical purity

enantiomeric ratio: ≥99.5:0.5 (HPLC)

quality

LiChropur

mp

127-131 °C (lit.)
127-131 °C

SMILES string

Cc1ccc2[N@H]3C[N@@H](Cc2c1)c4ccc(C)cc4C3

InChI

1S/C17H18N2/c1-12-3-5-16-14(7-12)9-18-11-19(16)10-15-8-13(2)4-6-17(15)18/h3-8H,9-11H2,1-2H3

InChI key

SXPSZIHEWFTLEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tröger base (TB) is a chiral heterocyclic amine with chirality due to the presence of two stereogenic nitrogen atoms.[1]

Application

It was used as an standard analyte for testing the enantioseparation capability of the chiral stationary phase column; in a study to understand the covalently bonded cellulose tris(3,5-dimethylphenylcarbamate) on a silica monolithic capillary column.[2]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Chiral diamine with hindered inversion at the nitrogen atoms[3]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V. Prelog et al.
Helvetica Chimica Acta, 27, 1127-1127 (1944)
Xiaoli Dong et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 875(1), 317-322 (2008-09-02)
A chiral capillary monolithic column for capillary electrochromatography (CEC) was prepared by covalent bonding of cellulose tris(3,5-dimethylphenylcarbamate) (CDMPC) on the silica monolithic matrix within the confine of a 50-microm i.d. bare fused silica capillary. Several pairs of enantiomers including neutral
Resolution, asymmetric transformation, and configuration of Troger's base. Application of Troger's base as a chiral solvating agent.
Wilen, Samuel H., Jian Zhong Qi, and Paul G. Williard
Organic Chemistry, 56, 485-487 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service