40642
Dimethyl sebacate
analytical standard
Synonym(s):
Decanedioic acid 1,10-dimethyl ester, Dimethyl decanedioate, NSC 9415, Sebacic acid dimethyl ester
About This Item
grade
analytical standard
Quality Level
assay
≥99.0% (GC)
shelf life
limited shelf life, expiry date on the label
bp
158 °C/10 mmHg (lit.)
mp
29-31 °C (lit.)
density
0.988 g/mL at 25 °C (lit.)
application(s)
environmental
SMILES string
COC(=O)CCCCCCCCC(=O)OC
InChI
1S/C12H22O4/c1-15-11(13)9-7-5-3-4-6-8-10-12(14)16-2/h3-10H2,1-2H3
InChI key
ALOUNLDAKADEEB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
signalword
Danger
hcodes
Hazard Classifications
Aquatic Chronic 2 - Eye Dam. 1
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
314.6 °F - closed cup
flash_point_c
157 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service