Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40585

Supelco

Cyanide Ionophore II

Selectophore, function tested

Synonym(s):

Magnesium phthalocyanine, Phthalocyanine magnesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H16MgN8
CAS Number:
Molecular Weight:
536.83
Beilstein/REAXYS Number:
3584903
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

quality

function tested

SMILES string

[Mg]1n2c3nc4nc(nc5n1c(nc6nc(nc2c7ccccc37)c8ccccc68)c9ccccc59)c%10ccccc4%10

InChI

1S/C32H16N8.Mg/c1-2-10-18-17(9-1)25-33-26(18)38-28-21-13-5-6-14-22(21)30(35-28)40-32-24-16-8-7-15-23(24)31(36-32)39-29-20-12-4-3-11-19(20)27(34-29)37-25;/h1-16H;/q-2;+2

InChI key

NIQREIWCURIAMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyanide Ionophore II (Magnesium phthalocyanine, MgPc) is a metal incorporated phthalocyaninato(2-) macro-ligand. This metallophthalocyanine has its absorption spectrum depending on the thickness of the film.[1]

Application

Preparation of membrane sensors for cyanide ions[2].

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X-ray single crystal investigations of magnesium phthalocyanine. The 4+ 1 coordination of the Mg ion and its consequence
Janczak, Jan, and Ryszard Kubiak
Polyhedron, 20, 2901-2909 (2001)
Hassan, S.S.M., et al.
Electroanalysis, 16, 298-303 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service